Radioactive Tape
Radioactive Tape
Actual product may vary
Fisherbrand
Radioactive Tape
Apply to bottles, boxes, and vials—anything that may be a radioactive material. Easy to write on.
Have Questions?
Catalog NumberDisplay Text
FIS15-943Radiation symbol, caution and printed headings
Catalog number FIS15-943
Price (MYR)
21.00
Add to cart
Display Text:
Radiation symbol, caution and printed headings
Price (MYR)
21.00
Add to cart
Apply to bottles, boxes, and vials—anything that may be a radioactive material. Easy to write on.
  • Yellow tape with bright red imprint with radiation symbol and “CAUTION RADIOACTIVE MATERIAL” message
  • Also available with printed headings for Isotope, Amount, and Date followed by space for notations to identify specific material
Specifications
ColorYellow
FormatTape
Product TypeRadioactive Tape
Display TextRadiation symbol, caution and printed headings
For Use With (Application)Bottles, boxes and vials anything that may be a radioactive material
Quantity1 Ea.
Dimensions (LxW)14 yd. x 0.75 in.
Length (English)14 yd.
Length (Metric)13 m
Width (English)0.75 in.
Unit Size