Lab Hazard Warning Label Refills
Lab Hazard Warning Label Refills
Actual product may vary
Fisherbrand
Lab Hazard Warning Label Refills
Printed in safety colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDisplay Text
FIS15-933Corrosive
FIS15-934Flammable
Catalog number FIS15-933
Price (MYR)
16.00
Add to cart
Display Text:
Corrosive
Price (MYR)
16.00
Add to cart
Printed in safety colors.
For use with Fisherbrand™ Lab Hazard Warning Label Dispenser (cat. no. 15931).
  • Tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll
Specifications
ColorAssorted
FormatTape
Product TypeCorrosive Warning Refill Tape
Display TextCorrosive
For Use With (Application)Printed in safety colors
Quantity4/Cs.
Length (English)57 yd.
Length (Metric)20.3 m
Width (English)0.5 in.
Unit Size