Check Order Status
Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles
Actual product may vary
Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles
Have Questions?
Catalog Number
Length (Metric) Sleeve
Size
Stopper Number
FIS14-126BB
1.5cm
14.5
14.5
Catalog number
FIS14-126BB
Price (MYR)
921.00
PK
Add to cart
Length (Metric) Sleeve:
1.5cm
Size:
14.5
Stopper Number:
14.5
Price (MYR)
921.00
PK
Add to cart
Comprised of dry natural rubber
Folds down over neck of bottle
With hollow plugs, rounded corners and needle guide
Easily perforated with hypodermic needles
Self-sealing
Specifications
Diameter (Metric) Plug
1.38cm
For Use With (Equipment)
Serum and vaccine bottles
Length (Metric) Sleeve
1.5cm
Size
14.5
Color
Red
Material
Rubber
Product Type
Sleeve Stopper
Stopper Number
14.5