Cast-Iron Rings
Cast-Iron Rings
Actual product may vary
Fisherbrand
Cast-Iron Rings
Cast-iron construction prevents breakage under normal use
Have Questions?
Catalog NumberDiameter (English) InnerDiameter (English) OuterDiameter (Metric) InnerDiameter (Metric) Outer
FIS14-050BQ2.37 in.3 in.6 cm7.6 cm
Catalog number FIS14-050BQ
Price (MYR)
-
Diameter (English) Inner:
2.37 in.
Diameter (English) Outer:
3 in.
Diameter (Metric) Inner:
6 cm
Diameter (Metric) Outer:
7.6 cm
Cast-iron construction prevents breakage under normal use
  • Sturdy 0.01 in. (7.9mm) setscrew in integral clamp locks ring tightly to support stands, lattices, etc.
  • Ring center is standard 4.5 in. (11cm) from support rod center
  • 2 in. (51mm) Size ring has lugs for holding crucibles
Specifications
MaterialCast Iron
Diameter (English) Inner2.37 in.
Diameter (English) Outer3 in.
Diameter (Metric) Inner6 cm
Diameter (Metric) Outer7.6 cm
Product TypeCast Iron Ring
ShapeRound
For Use With (Application)Prevents breakage under normal use
Quantity1 Ea.
Unit Size