Glass Ubbelohde Calibrated Viscometer Tubes
Glass Ubbelohde Calibrated Viscometer Tubes
Actual product may vary
Fisherbrand
Glass Ubbelohde Calibrated Viscometer Tubes
For measurement of kinematic viscosities of petroleum products according to ASTM™ D 445 and D 446.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberApproximate ConstantASTM NumbersSizeViscosity Range
FIS13-614A0.01 cSt/sec.112 to 10 cSt
FIS13-614F0.003 cSt/sec.0C0C0.6 to 3 cSt
Catalog number FIS13-614A
Price (MYR)
2,801.00
Add to cart
Approximate Constant:
0.01 cSt/sec.
ASTM Numbers:
1
Size:
1
Viscosity Range:
2 to 10 cSt
Price (MYR)
2,801.00
Add to cart
For measurement of kinematic viscosities of petroleum products according to ASTM™ D 445 and D 446.
  • Certified accurate (certificate included)
  • Excellent precision
  • Borosilicate glass
  • Minimum sample volume: 11mL
  • Require liquid bath depth of 241mm
Specifications
Product TypeUbbelohde Viscometer
Viscosity Range2 to 10 cSt
CalibratedYes
Approximate Constant0.01 cSt/sec.
ASTM Numbers1
Certifications/ComplianceASTM D445, ASTM D446, ISO 3104, ISO 3105, calibration certificate
IncludesInstruction sheet, calibration certificate
MaterialBorosilicate Glass
Depth (English) Bath9.5 in.
Depth (Metric) Bath24.1 cm
AccuracyASTM D445, ASTM D446
Precision±0.2%
For Use With (Application)For measurement of kinematic viscosity of transparent Newtonian liquids
Size1
Volume (Metric)11 mL
Unit Size