Stainless-Steel Utility Trays
Stainless-Steel Utility Trays
Actual product may vary
Fisherbrand
Stainless-Steel Utility Trays
Corrosion resistant and easy to clean
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)Type
FIS13-361C16.5 x 10 x 2.5 in.Instrument Tray
FIS13-361B12.25 x 7.75 x 2.25 in.Instrument Tray
FIS13-361D20.75 x 12.75 x 2.5 in.Catheter Tray
Catalog number FIS13-361C
Price (MYR)
469.00
Add to cart
Dimensions (L x W x H):
16.5 x 10 x 2.5 in.
Type:
Instrument Tray
Price (MYR)
469.00
Add to cart
Corrosion resistant and easy to clean
Specifications
Capacity (Metric)4.7 L
Dimensions (L x W x H)16.5 x 10 x 2.5 in.
Height (Metric)6.4 cm
Length (Metric)42 cm
MaterialStainless Steel
TypeInstrument Tray
Width (Metric)25 cm
IncludesSquare corners
Length (English)16.375 in.
Height (English)2.5 in.
ShapeRectangular
Width (English)9.75 in.
Capacity (English)5 qt.
Unit Size