Check Order Status
Plain Glass Microslides
Actual product may vary
Fisherbrand
Plain Glass Microslides
Individually selected for consistent quality.
Have Questions?
Change view
Catalog Number
Width (Metric)
FIS12-550C
50 mm
FIS12-550D
25 mm
2 Options
Catalog number
FIS12-550C
Price (MYR)
862.00
Add to cart
Width (Metric):
50 mm
Price (MYR)
862.00
Add to cart
Individually selected for consistent quality
Clear, corrosion-resistant glass with flat surfaces and ground edges
Precleaned and ready to use in one-at-a-time dispenser boxes of 72 slides
Specifications
Edges
Ground
Frosted
Not Frosted
Length (Metric)
75 mm
Material
Glass
Precleaned
Yes
Thickness (Metric)
1 mm
Type
Microscope Slide
Width (Metric)
50 mm
Color
Clear
Coating
Chemical Resistant
Unit Size