Brix Hydrometers
Brix Hydrometers
Actual product may vary
Fisherbrand
Brix Hydrometers
Used to measure the concentration of sugar solutions in refineries.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)For Use With (Equipment)Brix RangeIncrements
FIS11-608C300 mm300mL Borosilicate Glass cylinder (08-530J)19 to 31°0.1
FIS11-608D300 mm300mL Borosilicate Glass cylinder (08-530J)29 to 41°0.1°
FIS11-608A300 mm300mL Borosilicate Glass cylinder (08-530J)0 to 12°0.1
FIS11-608B300 mm300mL Borosilicate Glass cylinder (08-530J)9 to 21°0.1
Catalog number FIS11-608C
Price (MYR)
166.00
Add to cart
Length (Metric):
300 mm
For Use With (Equipment):
300mL Borosilicate Glass cylinder (08-530J)
Brix Range:
19 to 31°
Increments:
0.1
Price (MYR)
166.00
Add to cart
  • Calibrated at 20°C
  • Permanently marked with individual serial numbers
For measuring the concentration of sugar solutions in refineries
Specifications
Increments0.1
Certifications/ComplianceNIST, ASTM E100, ASTM E126
Length (Metric)300 mm
For Use With (Equipment)300mL Borosilicate Glass cylinder (08-530J)
Brix Range19 to 31°
MaterialSoda Lime Glass
For Use With (Application)Measuring the concentration of Sugar solutions in refineries
Volume (Metric)250 mL
Unit Size