Specific Gravity Hydrometers
Specific Gravity Hydrometers
Actual product may vary
Fisherbrand
Specific Gravity Hydrometers
Measure true specific gravity of liquids.
Have Questions?
Catalog NumberCertifications/ComplianceIncrementsLength (Metric)Specific Gravity Range
FIS11-520CASTM E100, ASTM E1260.002285 mm1.400 to 1.620
Catalog number FIS11-520C
Price (MYR)
159.00
Add to cart
Certifications/Compliance:
ASTM E100, ASTM E126
Increments:
0.002
Length (Metric):
285 mm
Specific Gravity Range:
1.400 to 1.620
Price (MYR)
159.00
Add to cart
Measure true specific gravity of liquids.
  • Chemical-resistant bulbs and stems
  • Streamlined to avoid trapping air bubbles
  • With directional device that stops the instrument with the scale facing the user for more convenient, more accurate readings
  • Double scales can be read without turning the hydrometer.
Specifications
TypeSingle Scale Hydrometer
Increments0.002
Certifications/ComplianceASTM E100, ASTM E126
Length (Metric)285 mm
For Use With (Equipment)300mL Borosilicate Glass cylinder
MaterialSoda Lime Glass
For Use With (Application)Measure true specific gravity of liquids heavier than Water
Specific Gravity Range1.400 to 1.620
Volume (Metric)250 mL
Unit Size