Flask Covers
Flask Covers
Actual product may vary
Flask Covers
Cut down on evaporation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure Size
FIS10-042B7 - 8
FIS10-042A6
Catalog number FIS10-042B
Price (MYR)
656.00
PK
Add to cart
Closure Size:
7 - 8
Price (MYR)
656.00
PK
Add to cart
  • Flint glass
  • Designed by Professor F.E. Tuttle
  • Place in necks of flasks while solutions are being boiled to cause condensation and refluxing
  • Use with any flask that takes rubber stopper sizes listed in ordering table
Specifications
Closure MaterialFlint Glass
MaterialGlass
For Use With (Equipment)Use with flask that takes Rubber stopper sizes, used when solutions are being boiled to cause condensation and refluxing
Product TypeCover, Flask
Closure Size7 - 8
Unit SizePK