Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Diameter (Metric) | Height (Metric) |
|---|---|---|---|
| FIS08-741D | |||
| CRG3140-100 | 325 mL | 100 mm | 50 mm |
| FIS08-741F | |||
| CRG3140-190 | 2500 mL | 190 mm | 100 mm |
| FIS08-741H | 2500 mL | 190 mm | 100 mm |
The rims on these dishes are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization (wet or dry). Ideal for storage and crystallization.