Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Actual product may vary
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Fisherbrand
Standard Class B Graduated Cylinders
Calibrated To Deliver
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsTolerance MetricSubdivision
FIS08-550A5 mL0.1 mL±0.1 mL0.1mL
FIS08-550B10 mL0.2 mL±0.2 mL0.2mL
FIS08-550D50 mL1 mL±0.5 mL1mL
FIS08-550C25 mL0.5 mL±0.34 mL0.5mL
FIS08-550G500 mL5 mL±4 mL5mL
FIS08-550E100 mL1 mL±1 mL1mL
Catalog number FIS08-550A
Price (MYR)
138.00
Add to cart
Capacity (Metric):
5 mL
Graduations:
0.1 mL
Tolerance Metric:
±0.1 mL
Subdivision:
0.1mL
Price (MYR)
138.00
Add to cart
Calibrated To Deliver
  • White-ceramic graduations
  • Heavy round bases
  • Handy pouring spouts
  • Accuracy is assured even at the lowest graduations; first 10% volume is not graduated
Specifications
Base TypeRound
CalibrationTo Deliver
Scale ColorWhite
ShapeRound
ClassB
Height (English)4.7 in.
Height (Metric)120 mm
Tolerance Metric±0.1 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)5 mL
For Use With (Application)Accuracy assured even at lowest graduations
IncludesHeavy Round Base
ScaleWhite ceramic graduations
Graduations0.1 mL
AutoclavableAutoclavable
Limit of Error+/- 0.10mL
Subdivision0.1mL
Unit Size