Cell Lifters
Cell Lifters
Actual product may vary
Cell Lifters
Cell Lifters
Fisherbrand
Cell Lifters
Disposable lifters quickly remove cell layers
Have Questions?
Catalog number FIS08-100-240
Price (MYR)
8.00
Special offer
Online exclusive
Ends: 30-Jul-2025
155.00
Save 147.00 (95%)
Add to cart
Price (MYR)
8.00
Special offer
Online exclusive
Ends: 30-Jul-2025
155.00
Save 147.00 (95%)
Add to cart
Disposable lifters quickly remove cell layers
The Fisherbrand Cell Lifter is used with dishes and cell culture clusters for quick removal of cell layers. The one-piece design and chiseled blade provide improved control while minimizing damage during the cell removal process.
  • Sterilized and individually wrapped in peel-open packaging
  • Polyethylene copolymer blade with angled and chiseled edge
  • Maintains even contact with growth area in culture dishes
Specifications
MaterialPolyethylene
SterilitySterile
For Use With (Application)Removal of cell layers
For Use With (Equipment)Dishes and cell culture clusters
DisposableSingle-use
Length (English) Blade0.7 in.
Length (English) Handle7 in.
Length (Metric) Blade2 cm
Length (Metric) Handle17.78 cm
EdgesAngled and Chiseled
Product TypeCell Lifter
Unit Size