Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply
Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply
Actual product may vary
Fisherbrand
Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Packaging Quantity
FIS06-666A8.2 x 4.4 in.286 Wipes/Box, 60 Boxes/Cs.
FIS06-6668.2 x 4.4 in.286 Wipes/Box, 30 Boxes/Cs.
FIS06-666-1A11.2 x 12.3 in.198 Wipes/Box, 15 Boxes/Cs.
FIS06-666C16.4 x 14.4 in.144 Wipes/Box, 15 Boxes/Cs.
Catalog number FIS06-666A
Price (MYR)
45.00
Add to cart
Dimensions (L x W):
8.2 x 4.4 in.
Packaging Quantity:
286 Wipes/Box, 60 Boxes/Cs.
Price (MYR)
45.00
Add to cart
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
  • Single ply, light-duty wiper that can handle a variety of delicate tasks
  • Extra low-lint wipers made from 100% virgin wood fibers
  • Soft, nonabrasive cellulose-fiber wipers won't scratch most delicate surfaces
  • Easily wipe up liquid and dust
  • Absorb many times their own weight
  • Handy one-at-a-time dispensing packs with special plastic guard to reduce airborne lint, keep contaminants out of box and reduce electrostatic discharge
Specifications
Product TypeDelicate Task Wipe
Length (English)8.2 in.
Length (Metric)20.83 cm
Width (English)4.4 in.
Width (Metric)11.17 cm
MaterialCellulose
ColorWhite
DisposableSingle-use
Dimensions (L x W)8.2 x 4.4 in.
For Use With (Application)General-Purpose
Packaging Quantity286 Wipes/Box, 60 Boxes/Cs.
Ply1-ply
Unit Size