Round Covered Lead Flask Rings
Round Covered Lead Flask Rings
Actual product may vary
Fisherbrand
Round Covered Lead Flask Rings
Stabilize Erlenmeyer flasks immersed in water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (Metric) InnerWeight (English)Weight (Metric)
FIS05-86962 in.5.1 cm1 lb.453.5 g
FIS05-86951.6 in.4.1 cm0.5 lb.226.7 g
FIS05-86972.2 in.5.6 cm1.5 lb.680.3 g
Catalog number FIS05-8696
Price (MYR)
240.00
Add to cart
Diameter (English) Inner:
2 in.
Diameter (Metric) Inner:
5.1 cm
Weight (English):
1 lb.
Weight (Metric):
453.5 g
Price (MYR)
240.00
Add to cart
Stabilize Erlenmeyer flasks immersed in water baths.
  • Reduces floating, breakage and chemical spills
  • Bright blue coating
  • Chemical-resistant
  • Will not scratch flasks
  • Available in four diameter/weight combinations
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBlue
HoldsFlasks (250 to 1000 mL)
MaterialLead
ShapeRound
ShelvesNone
StackableYes
SterilityNon-sterile
For Use With (Equipment)250 to 1000mL Erlenmeyer flasks immersed in water baths
Length (Metric)89 mm
Width (Metric)40 mm
Height (Metric)89 mm
Diameter (English) Inner2 in.
Diameter (Metric) Inner5.1 cm
Weight (English)1 lb.
Weight (Metric)453.5 g
Unit Size