Round Covered Lead Flask Rings
Round Covered Lead Flask Rings
Actual product may vary
Fisherbrand
Round Covered Lead Flask Rings
Stabilize Erlenmeyer flasks immersed in water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerDiameter (Metric) InnerWeight (English)Weight (Metric)
FIS05-86951.6 in.4.1 cm0.5 lb.226.7 g
FIS05-86962 in.5.1 cm1 lb.453.5 g
FIS05-86972.2 in.5.6 cm1.5 lb.680.3 g
Catalog number FIS05-8695
Price (MYR)
217.00
Add to cart
Diameter (English) Inner:
1.6 in.
Diameter (Metric) Inner:
4.1 cm
Weight (English):
0.5 lb.
Weight (Metric):
226.7 g
Price (MYR)
217.00
Add to cart
Stabilize Erlenmeyer flasks immersed in water baths.
  • Reduces floating, breakage and chemical spills
  • Bright blue coating
  • Chemical-resistant
  • Will not scratch flasks
  • Available in four diameter/weight combinations
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBlue
HoldsFlasks (125 to 500 mL)
MaterialLead
ShapeRound
ShelvesNone
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)125 to 500mL Erlenmeyer Flasks Immersed in Water Baths
Length (Metric)76 mm
Width (Metric)40 mm
Height (Metric)76 mm
Diameter (English) Inner1.6 in.
Diameter (Metric) Inner4.1 cm
Weight (English)0.5 lb.
Weight (Metric)226.7 g
Unit Size