Specifications
CalibrationTo contain
Certifications/ComplianceUSP, ASTM E 438, D2158
Capacity (Metric)100 mL
DisposableReusable
MaterialBorosilicate Glass
For Use WithUsed in the determination of residues in liquified petroleum (LP) gases
Tolerance0 to 0.1mL in ±0.02mL, >0.1 to 0.3mL in ±0.03mL, >0.3 to 1mL in ±0.05mL, >1 to 3mL in ±0.1mL, >3 to 5mL in ±0.2mL, >5 to 10mL in ±0.
ShapeConical
Length (Metric)203 mm
Bottom ShapeConical
Diameter (Metric)37 mm
Graduations0 to 1 mL in 0.05 mL increments, 1 to 3 mL in 0.1 mL increments, 3 to 6 mL in 0.2 mL increments, 6 to 10 mL in 0.5 mL incrementse, 10 to 100 mL in 1 mL increments
Product TypeCentrifuge Tube
Max. RCF800 x g
Unit Size