Bad Mouse anti-Human, Mouse, Clone: BYC001, MilliporeSigma™
Bad Mouse anti-Human, Mouse, Clone: BYC001, MilliporeSigma™
Actual product may vary
DWK Life Sciences
Bad Mouse anti-Human, Mouse, Clone: BYC001, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-605
Price (MYR)
4,160.00
Add to cart
Price (MYR)
4,160.00
Add to cart
This Anti-Bad Antibody, clone BYC001 is validated for use in immunoprecipitation, western blotting for the detection of Bad.
For Research Use Only
Specifications
AntigenBad
ApplicationsImmunoprecipitation, Western Blot
ClassificationMonoclonal
CloneBYC001
ConjugateUnconjugated
Formulation0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide.
Gene Accession No.NM_032989
Gene SymbolsBAD, BBC2, BBC6, BCL2L8
Host SpeciesMouse
ImmunogenFull-length murine Bad containing a N-terminal His6-tag
Purification MethodProtein G chromatography
Quantity200 μg
Primary or SecondaryPrimary
Target SpeciesHuman, Mouse
FormPurified
IsotypeIgG
Unit Size
Contents & Storage
2-8°C for one year