Phospho-STAT5A/B (Tyr694/699) Mouse anti-Human, Mouse, Rat, Bovine, Sheep, Clone: 8-5-2, MilliporeSigma™
Phospho-STAT5A/B (Tyr694/699) Mouse anti-Human, Mouse, Rat, Bovine, Sheep, Clone: 8-5-2, MilliporeSigma™
Actual product may vary
Phospho-STAT5A/B (Tyr694/699) Mouse anti-Human, Mouse, Rat, Bovine, Sheep, Clone: 8-5-2, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-495
Price (MYR)
207.00
EA
Add to cart
Price (MYR)
207.00
EA
Add to cart
Anti-phospho-STAT5A/B (Tyr694/699) Antibody, clone 8-5-2 is a mouse monoclonal antibody for detection of phospho-STAT5A/B (Tyr694/699) also known as signal transducer and activator of transcription 5A/B and has been validated in western blotting.
For Research Use Only
Specifications
Antigenphospho-STAT5A/B (Tyr694/699)
ApplicationsWestern Blot
ClassificationMonoclonal
Clone8/5/2002
ConjugateUnconjugated
FormulationPurified mouse monoclonal IgG in buffer containing PBS. Frozen solution at −20°C.
Gene Accession No.NM_003152.2
Gene SymbolsSTAT5A, MGF, STAT5
Host SpeciesMouse
ImmunogenKLH-conjugated, synthetic phosphopeptide (KAVDG[pY]VKPQIK) corresponding to amino acids 689-700 of STAT5A and 694-705 of STAT5B. Clone 8-5-2.
Purification MethodProtein G purfied
Quantity100 μg
Primary or SecondaryPrimary
Target SpeciesBovine, Human, Mouse, Rat, Sheep
FormPurified
IsotypeIgG
Unit SizeEA
Contents & Storage
Stable for 1 year at from date of receipt.