Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Length (Metric) | Diameter (Metric) |
---|---|---|---|
FIS033410 | 4 mL | 75 mm | 10 mm |
FIS03-341-0 | 4 mL | 75 mm | 10 mm |
FIS03-341-1 | 6 mL | 75 mm | 12 mm |
FIS033411 | 6 mL | 75 mm | 12 mm |
FIS033415 | 19 mL | 125 mm | 16 mm |
FIS03-341-3 | 10 mL | 100 mm | 13 mm |