General-Purpose Laboratory Brushes
Product Image
Fisherbrand
General-Purpose Laboratory Brushes
For dusting or painting.
Have Questions?
Catalog NumberColor
FIS03-645ABlack
Catalog number FIS03-645A
Price (MYR)
8.00
Add to cart
Color:
Black
Price (MYR)
8.00
Add to cart
For dusting or painting.
  • Tapered nylon bristles secured to plastic handle
Specifications
Product TypeBrush
Bristle MaterialNylon
Bristle TypeTapered
Handle MaterialPlastic
Tip TypeTapered
ColorBlack
Diameter (English) Bristle1 in.
Diameter (Metric) Bristle2.5 cm
For Use With (Application)For dusting or painting
Length (English)7.5 in.
Length (English) Bristle2 in.
Length (Metric)19 cm
Length (Metric) Bristle5.1 cm
SizeMedium
Unit Size