Flexible Brushes
Flexible Brushes
Actual product may vary
Fisher Catalogue
Flexible Brushes
Soft, single row of bristles fits easily into flask necks.
Have Questions?
Catalog NumberLength (English)For Use With (Equipment)
FIS03-570B14.17 in.500mL flasks
Catalog number FIS03-570B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
14.17 in.
For Use With (Equipment):
500mL flasks
Soft, single row of bristles fits easily into flask necks.
  • Plastic handle bends to allow bristles to fit into container
  • Handle has hanging ring
Specifications
Product TypeFlexible Brush
Bristle TypeSoft
Handle MaterialPlastic
Handle TypeFlexible
Diameter (English) Bristle1.4 in.
Diameter (Metric) Bristle3.8 cm
For Use With (Equipment)500mL flasks
IncludesHanging ring
Length (English)14.17 in.
Length (English) Bristle3.9 in.
Length (Metric)36 cm
Length (Metric) Bristle10 cm
No. of Bristle Rows1
Unit Size