Nalgene™ LDPE, Round Carboy with Spigot
Nalgene™ LDPE, Round Carboy with Spigot
Actual product may vary
Nalgene™ LDPE, Round Carboy with Spigot
Nalgene™ LDPE, Round Carboy with Spigot
Nalgene™ LDPE, Round Carboy with Spigot
Fisherbrand
Nalgene™ LDPE, Round Carboy with Spigot
For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-963B25 L
FKS1052808110 L
Catalog number FIS02-963B
Price (MYR)
2,760.00
Add to cart
Capacity (Metric):
25 L
Price (MYR)
2,760.00
Add to cart
Thermo Scientific™ Nalgene Round LDPE Carboys are the icon of lab water dispensing and storage. Place one at every sink and every work station to enhance your productivity. Carboys have a spigot boss molded in once piece with the carboy for convenience, strength and leak resistance. Suitable for collecting and dispensing distilled water, acids and reagents. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
  • Molded in graduations in 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles make lifting comfortable for four largest sizes
  • Molded in spigot boss provides strength and leak resistance
  • Leakproof†
  • Carboy and Carton are recyclable in many communities
  • Manufactured from non-toxic materials
Warning: Do not autoclave this carboy. For autoclavable carboys, choose a Nalgene Polypropylene, Polypropylene copolymer, or Polycarbonate carboy.
Ordering Information:
Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.
Includes:
Polypropylene spigot and screw closure with TPE gasket † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Nalgene rugged LDPE carboys are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE carboys and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE carboys.
Specifications
AutoclavableNot Autoclavable
Capacity (English)6.6 gal.
Capacity (Metric)25 L
Closure MaterialPolypropylene
Closure Size83B
ColorWhite (Opaque)
DescriptionRound Carboys w/Spigot
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer287 mm
For Use With (Application)Collect and dispense distilled water, acids and reagents
GraduationsMolded-in graduations in 1 gal. and 5 L increments
Height (Metric) With Closure594 mm
IncludesIntegral shoulder handles, Molded-in spigot boss
MaterialLDPE
Product TypeCarboy with Spigot
Unit Size