Autoclavable Waste Bags: Printed
Autoclavable Waste Bags: Printed
Actual product may vary
Fisherbrand
Autoclavable Waste Bags: Printed
Printed style.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Capacity (English)
FIS01-826BClear30 x 24 in. (76 x 61 cm)12 to 14 gal.
FIS01-826AClear24 x 12 in. (61 x 30 cm)2 to 3 gal.
FIS01-826CClear36 x 24 in. (91 x 61 cm)13 to 16 gal.
FIS01-826EClear48 x 37 in. (122 x 94 cm)44 gal.
FIS01-828ERed48 x 37 in. (122 x 94 cm)44 gal.
FIS01-828ARed12 x 8 in. (30 x 20 cm)1 to 2 gal.
FIS01-828BRed19 x 14 in. (48 x 36 cm)3 to 4 gal.
FIS01-828CRed23 x 19 in. (58 x 48 cm)7 to 10 gal.
FIS01-828DRed35 x 25 in. (89 x 64 cm)13 to 16 gal.
Catalog number FIS01-826B
Price (MYR)
683.00
Add to cart
Color:
Clear
Dimensions (L x W):
30 x 24 in. (76 x 61 cm)
Capacity (English):
12 to 14 gal.
Price (MYR)
683.00
Add to cart
Printed style bags
  • Maximum temperature, 140°C (285°F); for use at 134°C (274°F)
  • 2mil thick
  • Supplied with autoclavable elastic steam-venting closures
Specifications
AutoclavableAutoclavable
Capacity (English)12 to 14 gal.
Certifications/ComplianceOSHA, 29 CFR1910.1030
Closure TypeElastic
ColorClear
IncludesPrinted in English and Spanish
MaterialPolypropylene
Temperature Range (Metric)140°C
Thickness (English)2 mil
Dimensions (L x W)30 x 24 in. (76 x 61 cm)
Length (English)30 in.
Length (Metric)76 cm
Width (English)24 in.
Width (Metric)61 cm
Unit Size