Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Diameter (English) Outer | Height (English) |
|---|---|---|---|
| FIS01-215C | |||
| FIS01-215A | 1 mL | 0.41 in. | 2.63 in. |
| FIS01-215D | 10 mL | 0.74 in. | 4.21 in. |
| FIS01-215B | 2 mL | 0.47 in. | 2.95 in. |
Prescored to eliminate need for filing