IgG Fab Goat anti-Rabbit, HRP, Polyclonal, Southern Biotech™
IgG Fab Goat anti-Rabbit, HRP, Polyclonal, Southern Biotech™
Actual product may vary
IgG Fab Goat anti-Rabbit, HRP, Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
Have Questions?
Catalog number FIL404005
View Price:Sign in to see your account pricing. Need an account? Register with us today.
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
AntigenIgG Fab
ApplicationsELISA
ClassificationPolyclonal
ConcentrationLot specific
ConjugateHRP
FormulationSupplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added
Host SpeciesGoat
ImmunogenRabbit IgG
Purification MethodAffinity Purified
Quantity1.0 mL
Regulatory StatusRUO
Primary or SecondarySecondary
Target SpeciesRabbit
IsotypeIgG
Unit Size
Contents & Storage
Store at 2-8°C