Check Order Status
L-Threonine (White Crystals or Crystalline Powder), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
L-Threonine (White Crystals or Crystalline Powder), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FGCBP394-100
100 g
Catalog number
FGCBP394-100
Price (MYR)
486.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
486.00
EA
Add to cart
Specifications
Chemical Name or Material
L-Threonine
Melting Point
255°C
Ammonium
0.02% max.
CAS
72-19-5
ChemAlert Storage Symbol
Gray
View more
This amino acid is suitable for use in tissue culture systems requiring additives.