L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™
Fisher Chemical
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™
Have Questions?
Catalog NumberQuantity
FGCBP384-100100 g
Catalog number FGCBP384-100
Price (MYR)
586.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
586.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Isoleucine
Melting Point284°C
Ammonium0.02% max.
CAS73-32-5
ChemAlert Storage SymbolGray
View more
This amino acid is suitable for use in tissue culture systems requiring additives.