Nystatin, Fisher BioReagents
Nystatin, Fisher BioReagents
Nystatin, Fisher BioReagents
Nystatin, Fisher BioReagents
Fisher Chemical
Nystatin, Fisher BioReagents
Alters membrane permeability and allows potassium ion leakage
Have Questions?
Catalog NumberQuantity
FGCBP2949-55 g
Catalog number FGCBP2949-5
Price (MYR)
529.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
529.00
EA
Add to cart
Specifications
Chemical Name or MaterialNystatin
TypeNystatin
Physical FormSolid
CAS1400-61-9
Health Hazard 2CAUTION!
View more