Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
Actual product may vary
Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
Fisher Chemical
Ethidium Bromide (1% Solution/Molecular Biology), Fisher BioReagents
CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol
Have Questions?
Catalog NumberQuantity
FGCBP1302-1010 mL
Catalog number FGCBP1302-10
Price (MYR)
377.00
EA
Add to cart
Quantity:
10 mL
Price (MYR)
377.00
EA
Add to cart
Amber Poly Bottle; 10mL
Ethidium Bromide is an intercalating dye used in the visualization of DNA and RNA in electrophoresis gels and cesium chloride density gradients.
Specifications
Chemical Name or MaterialEthidium Bromide
ColorRed
Name Note1% Solution
CAS1239-45-8
ChemAlert Storage SymbolBlue
CAS7732-18-5
Recommended StorageRT
Purity Grade NotesDNase-, RNase- and Protease-Free
Molecular FormulaC21H20BrN3
MDL NumberMFCD00011724
Merck Index15, 4769
Synonymethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
InChI KeyZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
Molecular Weight (g/mol)394.32
PubChem CID14710
ChEBICHEBI:4883
Formula Weight394.33
GradeMolecular Biology
PackagingClear Bottle
Quantity10 mL
Physical FormLiquid
Unit SizeEA