Check Order Status
Toluidine Blue O, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Toluidine Blue O, Fisher BioReagents™
This nuclear stain may be used as an alternative to methylene blue
Have Questions?
Catalog Number
Quantity
FGCBP107-10
10 g
Catalog number
FGCBP107-10
Price (MYR)
633.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
633.00
EA
Add to cart
Specifications
Chemical Name or Material
Toluidine Blue O
CAS
92-31-9
Recommended Storage
RT
Extinction Coefficient
700g(-1) cm(-1) min.
Molecular Formula
C15H16ClN3S
View more