Check Order Status
Acetic Acid Glacial Min. 99%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetic Acid Glacial Min. 99%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Catalog number
FGCA507-P212
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
pH
2.5
CAS
64-19-7
Melting Point
16°C
Boiling Point
117°C
Color
Colorless
View more