Check Order Status
Hydrochloric Acid 32-35%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid 32-35%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
500 mL
Catalog number
FGCA466-1
Price (MYR)
6,064.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
6,064.00
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7647-01-0
Melting Point
-35°C
Boiling Point
57°C
View more