Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Fisher Chemical
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
4 L
Packaging:
View all
Glass Bottle
Catalog number FGCA461-212
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13822-56-5
IUPAC Name(3-aminopropyl)trimethoxysilane
Molecular FormulaC6H17NO3Si
InChI KeySJECZPVISLOESU-UHFFFAOYSA-N
SMILESCO[Si](CCCN)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Refractive index1.4220 to 1.4250 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm