Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
Fisher Chemical
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
4 L
Packaging:
View all
Wide Neck Amber Bottle
Catalog number FGCA461-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Wide Neck Amber Bottle
Request bulk or custom format
Chemical Identifiers
CAS112-24-3
IUPAC Name(2-aminoethyl)({2-[(2-aminoethyl)amino]ethyl})amine
Molecular FormulaC6H18N4
InChI KeyVILCJCGEZXAXTO-UHFFFAOYSA-N
SMILESNCCNCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Refractive index1.4960 to 1.5000 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=55 %
View more
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm