Check Order Status
Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 10), TCI America™
Element Search
Structure Search
Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 10), TCI America™
Have Questions?
Catalog Number
Quantity
FCSP070725G
25 g
Catalog number
FCSP070725G
Price (MYR)
148.51
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
148.51
Add to cart
Specifications
Chemical Name or Material
Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 10)
CAS
26027-38-3
Color
Yellow
RTECS Number
RB1299090
pH
6.7 (1% H
2
O soln. 25°C)
View more