Check Order Status
Water, Distilled, Reagent Grade, Ricca Chemical
Element Search
Structure Search
Water, Distilled, Reagent Grade, Ricca Chemical
Have Questions?
Change view
Quantity:
4 L
20 L
Packaging:
View all
Amber Glass Bottle
Catalog number
FCS9180-1
Price (MYR)
239.87
Add to cart
Quantity:
4 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
239.87
Add to cart
Specifications
Chemical Name or Material
Water
Shelf Life
18 months
Name Note
Distilled
CAS
7732-18-5
Color
Colorless
View more