EZH2 Polyclonal Antibody, Active Motif™
EZH2 Polyclonal Antibody, Active Motif™
Actual product may vary
EZH2 Polyclonal Antibody, Active Motif™
EZH2 Polyclonal Antibody, Active Motif™
EZH2 Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
Have Questions?
Catalog NumberQuantity
FCS3990-1100 μL
Catalog number FCS3990-1
Price (MYR)
548.88
Add to cart
Quantity:
100 μL
Price (MYR)
548.88
Add to cart
EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
Specifications
AntigenEZH2
ApplicationsChIP Assay, ChIP sequencing (ChIP-seq), Immunohistochemistry
ClassificationPolyclonal
ConcentrationConc. Not Determined
ConjugateUnconjugated
Formulationwhole serum with 30% glycerol and 0.035% sodium azide; pH
GeneEZH2
Gene Accession No.Q15910, Q61188
Gene Aliasan; enhancer of zeste 2 polycomb repressive complex 2 subunit; Enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; ENX-1; Enx1h; EZH 2; EZH1; Ezh2; EZH-2; EZH2b; Histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; Lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2
Gene SymbolsEZH2
Host SpeciesRabbit
ImmunogenThis EZH2 antibody was raised against a recombinant fusion protein corresponding to amino acids 1-370 of mouse EZH2.
Quantity100 μL
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)14056, 2146
Target SpeciesHuman, Mouse
Product TypeAntibody
FormLiquid
IsotypeIgG
Unit Size
Contents & Storage
-20°C, Avoid Freeze/Thaw Cycles