Contrad™ 70 Liquid Detergent
Contrad™ 70 Liquid Detergent
Actual product may vary
Fisherbrand
Contrad™ 70 Liquid Detergent
Cleans glass, plastic, ceramic, and ferrous metals by soaking, scrubbing, or ultrasonic cleaning
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)
FCS04-35534 oz.
FCS04-355-11.33 gal.
Catalog number FCS04-355
Price (MYR)
301.00
Add to cart
Volume (English):
34 oz.
Price (MYR)
301.00
Add to cart
Cleans glass, plastic, ceramic, and ferrous metals by soaking, scrubbing, or ultrasonic cleaning
Can safely replace acid washes and caustic baths. Excellent substitute for chromic acid.
  • Concentrated liquid
  • Removes proteins, grease, oil, and baked-on contaminants
  • Nontoxic, phosphate-free, chlorine-free, and completely free-rinsing
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Product TypeLiquid Detergent
Color-codedNo
FormConcentrated Liquid
pH13
ScentRegular Scent
SterilityNon-sterile
AppearanceWhite
Boiling Point100°C
Certifications/ComplianceUSDA, 29 CFR 1910.1200
Size1 L
Volume (English)34 oz.
Unit Size