Check Order Status
Xylenes (Histological), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Xylenes (Histological), Fisher Chemical™
Have Questions?
Change view
Quantity:
1 gal.
4 L
Packaging:
View all
Poly Bottle
Catalog number
FCMX3P-1GAL
Price (MYR)
449.00
Add to cart
Quantity:
1 gal.
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
449.00
Add to cart
Specifications
Chemical Name or Material
Xylenes
Cleanup Symbol
Solvent
Melting Point
-34°C
CAS
1330-20-7
ChemAlert Storage Symbol
Red
View more