Check Order Status
Water (HPLC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Water (HPLC), Fisher Chemical™
Have Questions?
Change view
Quantity:
4 L
1 L
Packaging:
View all
Glass Bottle
Catalog number
FCMW5-4
Price (MYR)
476.00
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
476.00
Add to cart
Specifications
Chemical Name or Material
Water
pH
7
CAS
7732-18-5
ChemAlert Storage Symbol
Gray
Color
Colorless
View more
Improved filtration! HPLC mobile phase for analytical and preparative separations.