Check Order Status
Tetrahydrofuran (HPLC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Tetrahydrofuran (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Have Questions?
Change view
Quantity:
4 L
1 L
Packaging:
View all
Glass Bottle
Catalog number
FCMT425-4
Price (MYR)
898.00
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
898.00
Add to cart
Chemical Identifiers
CAS
787-69-9
IUPAC Name
1-{4'-acetyl-[1,1'-biphenyl]-4-yl}ethan-1-one
Molecular Formula
C16H14O2
InChI Key
YSTSBXDVNKYPTR-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(C)=O
View more
Specifications
Specification Sheet
Appearance (Color)
Colorless to tan
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Melting point
192°C to 197°C