Check Order Status
Thymol Blue (Certified ACS), Fisher Chemical
Element Search
Structure Search
Thymol Blue (Certified ACS), Fisher Chemical
pH Indicator
Have Questions?
Catalog Number
Quantity
FCMT416-5
5 g
Catalog number
FCMT416-5
Price (MYR)
148.69
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
148.69
Add to cart
Specifications
Chemical Name or Material
Thymol Blue
Melting Point
222.8°C
CAS
76-61-9
ChemAlert Storage Symbol
Gray
Clarity
Pass Test
View more