Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Fisher Chemical
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical™
Alkalimetric standard
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
View all
Poly Bottle
Catalog number FCMT395-500
Price (MYR)
660.00
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
660.00
Add to cart
Specifications
AbsorbancePass Test
Chemical Name or MaterialTris(hydroxymethyl)aminomethane
CAS77-86-1
ChemAlert Storage SymbolGray
Melting Point168.5°C
View more