Check Order Status
Toluene (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Toluene (Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
1 L
500 mL
Packaging:
Glass Bottle
Catalog number
FCMT324-1
Price (MYR)
498.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
498.00
EA
Add to cart
Specifications
Chemical Name or Material
Toluene
Cleanup Symbol
Solvent
Melting Point
-95°C
CAS
108-88-3
ChemAlert Storage Symbol
Red
View more