Titanium (IV) Oxide Anhydrous (Powder/Certified), Fisher Chemical
Titanium (IV) Oxide Anhydrous (Powder/Certified), Fisher Chemical
Titanium (IV) Oxide Anhydrous (Powder/Certified), Fisher Chemical
Titanium (IV) Oxide Anhydrous (Powder/Certified), Fisher Chemical
Titanium (IV) Oxide Anhydrous (Powder/Certified), Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
FCMT315-500500 gPoly Bottle
Catalog number FCMT315-500
Price (MYR)
512.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
512.00
EA
Add to cart
Specifications
Boiling Point2900°C
Chemical Name or MaterialTitanium (IV) Oxide Anhydrous
Melting Point1855°C
CAS13463-67-7
ChemAlert Storage SymbolGray
View more