Starch Solution (Stabilized), Fisher Chemical™
Starch Solution (Stabilized), Fisher Chemical™
Starch Solution (Stabilized), Fisher Chemical™
Starch Solution (Stabilized), Fisher Chemical™
Fisher Chemical
Starch Solution (Stabilized), Fisher Chemical™
For iodometry
Have Questions?
Catalog NumberQuantityPackaging
FCMSS40811 LPoly Bottle
Catalog number FCMSS4081
Price (MYR)
399.00
Add to cart
Quantity:
1 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
399.00
Add to cart
Chemical Identifiers
CAS1317-61-9
IUPAC Namediiron(3+) λ²-iron(2+) tetraoxidandiide
Molecular FormulaFe3O4
InChI KeyWTFXARWRTYJXII-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Particle Size50-100nm
Appearance (Color)Black
FormPowder
pH6-8
View more
For iodometry