Check Order Status
Schiff Reagent Solution (Modified), Fisher Chemical
Element Search
Structure Search
Schiff Reagent Solution (Modified), Fisher Chemical
For detection of aldehydes
Have Questions?
Catalog Number
Quantity
FCMSS32-500
500 mL
Catalog number
FCMSS32-500
Price (MYR)
705.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
705.00
EA
Add to cart
Chemical Identifiers
CAS
632-99-5, 7681-57-4, 7647-01-0, 7732-18-5
Specifications
Chemical Name or Material
Schiff Reagent Solution
Name Note
Modified
CAS
632-99-5
ChemAlert Storage Symbol
White
CAS
7681-57-4
View more