Check Order Status
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
Actual product may vary
Fisher Chemical
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
Have Questions?
Change view
Catalog Number
Quantity
Packaging
FCMSP120-1
1 L
Glass Bottle
FCMSP120-500
500 mL
Glass Bottle
SP120-500
500 mL
Glass Bottle
3 Options
Catalog number
FCMSP120-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Specifications
Color
Yellow
pH Range
1.1
Format
Liquid
Packaging
Glass Bottle
Quantity
1 L
Type
Platinum Cobalt Color Standard Solution
Unit Size