Check Order Status
Ferric Chloride Solution, 0.025% w/v (APHA), Fisher Chemical
Element Search
Structure Search
Ferric Chloride Solution, 0.025% w/v (APHA), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMSF97-1
1 L
Glass Bottle
Catalog number
FCMSF97-1
Price (MYR)
487.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
487.00
EA
Add to cart
Specifications
Boiling Point
100°C
Chemical Name or Material
Ferric Chloride Solution, 0.025% (w/v)
Melting Point
0°C
CAS
7705-08-0
ChemAlert Storage Symbol
Gray
View more